Glycine ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
:75.07 g/mol
Merck Index:14,04491
Boiling Pt:181 °C (1013 hPa)
Melting Pt:240°C
Density:1,64 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-272-2
Specification Test Results
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Solubility in Water:almost transparency
- Drying loss:max. 0.2 %