5-Methylisatin ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:608-05-9
:
Formula:C₉H₇NO₂
MDL Number:MFCD00005721
:161.16 g/mol
EINECS:210-152-1
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:186.0 to 190.0 deg-C