Order Entry
Norway
ContactUsLinkComponent
2,2,4-Trimethylcyclopentanone ≥95.0% (by GC)
Flammable
2,2,4-Trimethylcyclopentanone ≥95.0% (by GC)
Catalog # TCIAT1719-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
2,2,4-Trimethylcyclopentanone ≥95.0% (by GC)
Catalog # TCIAT1719-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :126.2 g/mol
Boiling Pt:66°C
Flash Pt:38°C
EINECS:248-802-1
UN:1224
ADR:3,III
 :GHS02
CAS Number:28056-54-4
Formula:C₈H₁₄O
MDL Number:MFCD00019312