4-(Trifluoromethoxy)benzonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:332-25-2
:
Formula:C₈H₄F₃NO
MDL Number:MFCD00039474
Boiling Pt:193°C
Density:1,284 g/cm³ (20 °C)
Flash Pt:83°C
EINECS:206-363-3
UN:3276
ADR:6.1,III
:187.12 g/mol
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %