Order Entry
Norway
ContactUsLinkComponent
 
2-Bromo-α,α,α,4-tetrafluorotoluene ≥98.0% (by GC)
 :  TCI
 
undefined
2-Bromo-α,α,α,4-tetrafluorotoluene ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %