kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
3-Bromo-α,α,α,5-tetrafluorotoluene
3-Bromo-α,α,α,5-tetrafluorotoluene ≥98.0% (by GC)
3-Bromo-α,α,α,5-tetrafluorotoluene ≥98.0% (by GC)
:
TCI
3-Bromo-α,α,α,5-tetrafluorotoluene ≥98.0% (by GC)
:
TCI
:
130723-13-6
:
:
C₇H₃BrF₄
:
MFCD00042498
:
243 g/mol
:
GHS07
Appearance:
Colorless to Light yellow to Light orange clear liquid
Purity (GC):
min. 98.0 %
Manual Fluorination Reagents, Fluorinated Building Blocks