
3-Fluoro-o-toluidine ≥98.0% (by GC, titration analysis)
: TCI
:443-86-7
:
:C₇H₈FN
:MFCD00007760
:125.15 g/mol
:205°C
:7 °C
:1,359 g/cm³ (20 °C)
:86°C
:207-142-4
:6.1,III
:GHS07
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %