Order Entry
Norway
ContactUsLinkComponent
5-Chloro-o-toluidine ≥98.0% (by GC, titration analysis)
Irritant
Health
5-Chloro-o-toluidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAC1876-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
5-Chloro-o-toluidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAC1876-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to lump
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :141.6 g/mol
Boiling Pt:237°C
Melting Pt:22 °C
Density:1,17 g/cm³ (20 °C)
Flash Pt:160 °C
Storage Temperature:Ambient
EINECS:202-452-6
UN:2239
ADR:6.1,III
 :GHS07
 :GHS08
CAS Number:95-79-4
Formula:C₇H₈ClN
MDL Number:MFCD00007779