Order Entry
Norway
ContactUsLinkComponent
3-Chloro-o-toluidine ≥99.0% (by GC)
Irritant
Health
3-Chloro-o-toluidine ≥99.0% (by GC)
Catalog # TCIAC0300-25ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
3-Chloro-o-toluidine ≥99.0% (by GC)
Catalog # TCIAC0300-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Brown clear liquid
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :141.6 g/mol
Boiling Pt:245°C
Melting Pt:3°C
Density:1,185 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-756-6
UN:3429
ADR:6.1,III
 :GHS07
 :GHS08
CAS Number:87-60-5
Formula:C₇H₈ClN
MDL Number:MFCD00007766