
3-Bromo-p-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7745-91-7
:
Formula:C₇H₈BrN
MDL Number:MFCD00134176
:186.05 g/mol
Boiling Pt:139°C
Melting Pt:25…26 °C
Storage Temperature:Refrigerator
EINECS:231-807-8
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %