

5-Nitro-o-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:99-55-8
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007741
:152.15 g/mol
Melting Pt:108 °C
Density:1,365 g/cm³ (15 °C)
EINECS:202-765-8
UN:2660
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:107.0 to 111.0 deg-C
- Solubility in Methanol:within almost transparency