Order Entry
Norway
ContactUsLinkComponent
6-Chloro-m-toluidine ≥98.0% (by GC, titration analysis)
Toxic
Health
6-Chloro-m-toluidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAC2576-5G
Supplier:  TCI
CAS Number:  
undefined
Toxic
Health
6-Chloro-m-toluidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAC2576-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :141.6 g/mol
Boiling Pt:230°C
Melting Pt:30 °C
EINECS:202-454-7
UN:2239
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:95-81-8
Formula:C₇H₈ClN
MDL Number:MFCD00007674