



1,5-Hexadiene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:592-42-7
:
Formula:C₆H₁₀
MDL Number:MFCD00008666
:82.15 g/mol
Boiling Pt:59°C
Melting Pt:–141 °C
Density:0,692 g/cm³ (20 °C)
Flash Pt:–46°C
EINECS:209-754-7
UN:2458
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %