Order Entry
Norway
ContactUsLinkComponent
 
 
α,α',2,3,5,6-Hexachloro-p-xylene ≥98.0% (by GC)
 :  TCI
 
 
α,α',2,3,5,6-Hexachloro-p-xylene ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    176.0 to 182.0 deg-C
  • Solubility in Toluene:
    almost transparency