
Tetrafluoroisophthalonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2377-81-3
:
Formula:C₈F₄N₂
MDL Number:MFCD00013290
:200.1 g/mol
Boiling Pt:283 °C (1013 hPa)
Melting Pt:76…78 °C
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:75.0 to 79.0 deg-C