3-Aminophenol ≥98.5% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:591-27-5
:
Formula:C₆H₇NO
MDL Number:MFCD00007786
Merck Index:14,00460
Boiling Pt:164°C
Melting Pt:122 °C
Density:1,275 g/cm³ (20 °C)
Flash Pt:92 °C
UN:2512
ADR:6.1,III
:109.13 g/mol
EINECS:209-711-2
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.5 %
- Purity (Nonaqueous Titration):min. 98.5 %
- Melting point:121.0 to 125.0 deg-C
- Solubility in HCl (1+3):almost transparency