Triethyl citrate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:77-93-0
:
Formula:C₁₂H₂₀O₇
MDL Number:MFCD00009201
:276.29 g/mol
Merck Index:14,02326
Boiling Pt:294 °C (1013 hPa)
Melting Pt:–55 °C
Density:1,135 g/cm³ (25 °C)
Flash Pt:176°C
EINECS:201-070-7
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %