Ethyl butyrate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:105-54-4
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00009394
:116.16 g/mol
Merck Index:14,03775
Boiling Pt:120°C
Melting Pt:–92 °C
Density:0,88 g/cm³ (20 °C)
Flash Pt:23°C
Storage Temperature:Ambient
EINECS:203-306-4
UN:1180
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %