

1,4-Diamino-9,10-anthraquinone ≥90.0% (by HPLC, total nitrogen)
Catalog # TCIAD3539-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Gray to Dark purple to Black powder to crystal
- Purity (HPLC):min. 90.0 area%
- Purity (with Total Nitrogen):min. 90.0 %
Specification Test Results
About this item
:
:238.25 g/mol
Melting Pt:270°C
EINECS:204-922-6
:GHS07
:GHS08
CAS Number:128-95-0
Formula:C₁₄H₁₀N₂O₂
MDL Number:MFCD00001224