
2-Chloro-4-hydroxypyridine ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17368-12-6
:
Formula:C₅H₄ClNO
MDL Number:MFCD01646108
:129.55 g/mol
EINECS:241-399-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:168.0 to 173.0 deg-C