Order Entry
Norway
ContactUsLinkComponent
 
Heptadecanal ≥97.0% (by GC)
  TCIAH1295-1G
 :  TCI
 :  
undefined
Heptadecanal ≥97.0% (by GC)
  TCIAH1295-1G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    34.0 to 39.0 deg-C

 

 

 :
 :254.46 g/mol
 :318°C
 :211-115-2
 :GHS07
 :Refrigerator
 :629-90-3
 :C₁₇H₃₄O
 :MFCD04034658