Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2-methylpyridine
3-Bromo-2-methylpyridine ≥98.0% (by GC)
3-Bromo-2-methylpyridine ≥98.0% (by GC)
:
TCI
3-Bromo-2-methylpyridine ≥98.0% (by GC)
:
TCI
:
38749-79-0
:
:
C₆H₆BrN
:
MFCD00191224
:
172.02 g/mol
:
76°C
:
79°C
:
GHS07
Appearance:
Colorless to Light orange to Yellow clear liquid
Purity (GC):
min. 98.0 %
Manual Bioscience and Analyticalscience