Order Entry
Norway
ContactUsLinkComponent
Diallyl disulphide ≥80.0% (by GC)
Diallyl disulphide ≥80.0% (by GC)
Catalog # TCIAD0071-25G
Supplier:  TCI
CAS Number:  
undefined
Diallyl disulphide ≥80.0% (by GC)
Catalog # TCIAD0071-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 80.0 %

Specification Test Results

About this item

 :
 :146.28 g/mol
Boiling Pt:198 °C (1013 hPa)
Density:1,0237 g/cm³ (15 °C)
Flash Pt:62°C
Storage Temperature:Refrigerator
EINECS:218-548-6
UN:2810
ADR:6.1,III
 :GHS06
CAS Number:2179-57-9
Formula:C₆H₁₀S₂
MDL Number:MFCD00008656