

Piperazine hexahydrate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:142-63-2
:
Formula:C₄H₁₀N₂·6H₂O
MDL Number:MFCD00149389
:194.23 g/mol
Merck Index:14,07464
Boiling Pt:145 °C (1013 hPa)
Melting Pt:44…45 °C
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:203-808-3
UN:2579
ADR:8,III
:GHS05
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Melting point:42.0 to 46.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together