2-Cyclohexylcyclohexanol cis- and trans-mixture ≥98.0% (by GC)
TCIAC0498-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
:
:182.31 g/mol
:264°C
:229-430-9
:6531-86-8
:C₁₂H₂₂O
:MFCD00021278