Order Entry
Norway
ContactUsLinkComponent
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥98.0% (by total nitrogen basis)
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥98.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
undefined
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥98.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

Related Information