

2-Aminophenol ≥99.0% (by GC)
: TCI
:95-55-6
:
:C₆H₇NO
:MFCD00007690
:14,00461
:235 °C (1013 hPa)
:1,328 g/cm³ (20 °C)
:≥ 175 °C
:202-431-1
:6.1,III
:109.13 g/mol
:2512
:GHS07
:GHS08
- Appearance:White to Brown powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in dilute HCl:within almost transparency