Iodoacetamide ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:144-48-9
:
Formula:ICH₂CONH₂
MDL Number:MFCD00008028
:184.96 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:91…94 °C
Storage Temperature:Refrigerator
EINECS:205-630-1
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:93.0 to 97.0 deg-C
- Solubility in Methanol:almost transparency