3-Iodo-o-xylene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:31599-60-7
:
Formula:C₈H₉I
MDL Number:MFCD00041437
:232.06 g/mol
Boiling Pt:231°C
Density:1,64 g/cm³ (20 °C)
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 97.0 %