Bis(2-ethylhexyl)sebacate ≥98.0% (by GC)
: TCI
:122-62-3
:
:C₂₆H₅₀O₄
:MFCD00009497
:426.68 g/mol
:14,01251
:377°C
:–55 °C
:0,914 g/cm³ (20 °C)
:222°C
:Ambient
:204-558-8
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %