
Salicylamide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:65-45-2
:
Formula:C₇H₇NO₂
MDL Number:MFCD00007978
:137.14 g/mol
Merck Index:14,08328
Boiling Pt:182°C
Melting Pt:137 °C
Density:1,33 g/cm³ (20 °C)
EINECS:200-609-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:140.0 to 143.0 deg-C
- Solubility in Methanol:almost transparency