Order Entry
Norway
ContactUsLinkComponent
4-Amino-2,6-dichlorophenol ≥98.0% (by titrimetric analysis)
Irritant
4-Amino-2,6-dichlorophenol ≥98.0% (by titrimetric analysis)
Catalog # TCIAA0878-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-Amino-2,6-dichlorophenol ≥98.0% (by titrimetric analysis)
Catalog # TCIAA0878-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :178.02 g/mol
Boiling Pt:304 °C (1013 hPa)
EINECS:227-671-4
 :GHS07
CAS Number:5930-28-9
Formula:C₆H₅Cl₂NO
MDL Number:MFCD00007875