
4,4'-Dibromobenzil ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:35578-47-3
:
Formula:C₁₄H₈Br₂O₂
MDL Number:MFCD00000104
:368.02 g/mol
EINECS:252-628-1
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:224.0 to 229.0 deg-C