9-Bromophenanthrene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:573-17-1
:
Formula:C₁₄H₉Br
MDL Number:MFCD00001174
Boiling Pt:225°C
Melting Pt:62 °C
Density:1,4093 g/cm³ (101 °C)
:257.13 g/mol
Storage Temperature:Ambient
EINECS:209-351-6
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:62.0 to 67.0 deg-C
- Solubility in Toluene:almost transparency