Acetohydroxamic acid
Catalog # MOLEM22956976
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:75.0666 g/mol
EINECS:208-913-8
CAS Number:546-88-3
Formula:C₂H₅NO₂
MDL Number:MFCD00009994