Order Entry
Norway
ContactUsLinkComponent
(S)-(+)-3-Hydroxytetrahydrofuran 99%
Irritant
(S)-(+)-3-Hydroxytetrahydrofuran 99%
Certificates
undefined
Irritant
(S)-(+)-3-Hydroxytetrahydrofuran 99%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4490 to 1.4510 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific optical rotation:
    +15° to +20° (20°C, 589 nm) (c=2, MeOH)

Specification Test Results

Related Information