Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Carmine
Carmine, red powder C.I. 75470
Carmine, red powder C.I. 75470
ICNA0215263425
:
MP Biomedicals
:
Carmine, red powder C.I. 75470
ICNA0215263425
:
MP Biomedicals
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
492.389 g/mol
:
215-724-4
:
1390-65-4
:
MFCD00167031
:
C₂₂H₂₀O₁₃
:
MFCD00167028