Anti-static harnesses, Miller® ATEX
This harness is the perfect solution for work at height in explosive atmospheres. It is particularly suitable for petrochemical and mining environments. The anti-static material prevents the risks of an electrostatic discharge igniting the explosive atmosphere thus allowing the user to work in complete safety in these high-risk environments.
- With two anchorage points (rear and front): Versatile
- Shoulder and leg straps made of 45 mm polyester webbing (anti-static) with steel threads in the webbing to ensure total conductibility
- Dorsal extension strap facilitates connection to the intermediate attachment (lanyard or fall arrest block)
- Detachable dorsal pad facilitates harness donning and prevents the straps from tangling up
: EN 361, ATEX 949/CE, EN 13464-1, EN 13463-1