kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Pyridoxal 5'-phosphate monohydrate
Pyridoxal-5'-phosphate monohydrate
Pyridoxal-5'-phosphate monohydrate
Pyridoxal-5'-phosphate monohydrate
Supplier:
RESEARCH ORGANICS
Certificates
Expand image
Pyridoxal-5'-phosphate monohydrate
Supplier:
RESEARCH ORGANICS
About this item
CAS Number:
41468-25-1
:
Formula:
C₈H₁₀NO₆P·1H₂O
MDL Number:
MFCD00149414
:
265.17 g/mol (247.15 anhy)
Storage Temperature:
Freezer