Benzo[k]fluoranthene
Catalog # PROOCILULM24170.1
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
EINECS:205-916-6
CAS Number:207-08-9
Formula:C₂₀H₁₂
MDL Number:MFCD00046287