Order Entry
Norway
ContactUsLinkComponent
Caspase 3 Substrate IV (Z-DEVD-AFC, Fluorogenic)
Caspase 3 Substrate IV (Z-DEVD-AFC, Fluorogenic)
Catalog # 264150-1
CAS Number:  
Caspase 3 Substrate IV (Z-DEVD-AFC, Fluorogenic)
Catalog # 264150-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 mg
  • CAS No.:
    1177935-21-5
  • Storage conditions:
    Freezer
  • Protein/peptide name:
    Caspase 3 Substrate IV (Z-DEVD-AFC, Fluorogenic)
  • Molecular formula:
    C₃₆H₃₈F₃N₅O₁₅

Specifications

About this item