Dibutyl phthalate-[ring-D₄]
Catalog # EHERC16171010
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
Storage Temperature:Ambient
UN:3082
ADR:9,III
:GHS08
:GHS09
CAS Number:93952-11-5
Formula:C₁₆H₁₈D₄O₄