Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, chrysene 200 µg/ml in toluene, CIL
Organic reference standard, chrysene 200 µg/ml in toluene, CIL
PROOCILULM74241.2
:
LGC Standards PROMOCHEM
:
Organic reference standard, chrysene 200 µg/ml in toluene, CIL
PROOCILULM74241.2
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,2 mL
CAS No.:
218-01-9
Description:
Chrysene 200 µg/ml in toluene
Formula:
C₁₈H₁₂
Description:
Chrysene