
2-Fluoro-6-methylbenzaldehyde ≥97%
Catalog # H55228.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:138.14 g/mol
Density:1.151
:GHS07
Storage Temperature:Ambient
CAS Number:117752-04-2
Formula:C₈H₇FO
MDL Number:MFCD09258962