Order Entry
Norway
ContactUsLinkComponent
 
Pancreatin, Protease, Lipase, Amylase
  1.07133.5000
 :  
Pancreatin, Protease, Lipase, Amylase
  1.07133.5000
 :  

 

  • Fp:
    5 kg
  • CAS-nr:
    8049-47-6
  • Seq:
    451
  • Lagringsforhold:
    Refrigerator
  • Enzymnavn:
    Pancreatin;Protease;Lipase;Amylase
  • Nr.:
    Ph. Eur., 4 x USP
  • Uttestet applikasjon:
    Suitable for use as excipient

 

 

Pancreatin 1400 FIP-U/g Protease, 24000 FIP-U/g Lipase, 30000 FIP-U/g Amylase EMPROVE® exp Ph. Eur., USP, ESSENTIAL