Pararosaniline hydrochloride
Catalog # EHERC10425000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:323.82 g/mol
Melting Pt:268 °C
EINECS:209-321-2
:GHS08
Storage Temperature:Ambient
CAS Number:569-61-9
Formula:C₁₉H₁₈ClN₃
MDL Number:MFCD00001657