Order Entry
Norway
ContactUsLinkComponent
 
Whole blood genomic DNA purification mini kit, GeneJET™
 
undefined
Whole blood genomic DNA purification mini kit, GeneJET™
 

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    GeneJET™ Whole Blood Genomic DNA Purification Mini Kit