Cryoboxes, Cardboard
Cryo resistant cardboard.
- Use with cardboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
For storage of cryovials at low temperatures.