Nystatin
Catalog # ICNA194534.5
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 KU
- Seq:6190
Specifications
About this item
:
:926.09 g/mol
EINECS:215-749-0
:GHS07
Storage Temperature:Freezer
CAS Number:1400-61-9
Formula:C₄₇H₇₅NO₁₇
MDL Number:MFCD25541651