Nystatin
Catalog # EHERC15661000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:926.09 g/mol
EINECS:215-749-0
:GHS07
Storage Temperature:Freezer
CAS Number:1400-61-9
Formula:C₄₇H₇₅NO₁₇
MDL Number:MFCD25541651